Assay Details
| Gene Symbol: MGC109340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024267.2 | NP_001019438.1 | 1 | 1419 | 89 | |
| GenBank mRNA | BC092634.1 | - | 1 | 1421 | 7315105 | 89 |
More Information
Set Membership: |
|

MGC109340
Genomic Map