Assay Details
| Gene Symbol: Slfnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024347.1 | NP_001019518.1 | 4 - 5 | 1480 | 70 | |
| XM_008764062.1 | XP_008762284.1 | 5 - 6 | 3014 | 70 | ||
| GenBank mRNA | BC079280.1 | - | 4 - 5 | 1480 | 7134187 | 70 |
More Information
Set Membership: |
|

Slfnl1
Genomic Map