Assay Details
| Gene Symbol: Fam107b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025034.1 | NP_001020205.1 | 1 - 2 | 135 | 92 | |
| GenBank mRNA | BC085116.1 | - | 1 - 2 | 135 | 7107468 | 92 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085116.1, NM_001025034.1 transcripts.
Set Membership: |
|

Fam107b
Genomic Map