Assay Details
| Gene Symbol: Fam107b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025034.1 | NP_001020205.1 | 2 - 3 | 317 | 105 | |
| XM_008771879.1 | XP_008770101.1 | 2 - 3 | 716 | 105 | ||
| XM_008771880.1 | XP_008770102.1 | 2 - 3 | 339 | 105 | ||
| XM_008771881.1 | XP_008770103.1 | 2 - 3 | 765 | 105 | ||
| XM_008771882.1 | XP_008770104.1 | 2 - 3 | 352 | 105 | ||
| XM_008771883.1 | XP_008770105.1 | 2 - 3 | 969 | 105 | ||
| XM_008771884.1 | XP_008770106.1 | 2 - 3 | 361 | 105 | ||
| GenBank mRNA | BC085116.1 | - | 2 - 3 | 317 | 7107468 | 105 |
More Information
Set Membership: |
|

Fam107b
Genomic Map