Assay Details
| Gene Symbol: Iqcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025153.1 | NP_001020324.1 | 7 - 8 | 645 | 73 | |
| XM_006243837.1 | XP_006243899.1 | 7 - 8 | 681 | 73 | ||
| GenBank mRNA | BC079282.1 | - | 7 - 8 | 645 | 7134209 | 73 |
More Information
Set Membership: |
|

Iqcf3
Genomic Map