Assay Details
| Gene Symbol: Cox6b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039085.1 | NP_001034174.1 | 2 - 3 | 192 | 68 | |
| XM_008758979.1 | XP_008757201.1 | 2 - 3 | 325 | 68 | ||
| GenBank mRNA | AY152399.1 | - | 2 - 3 | 192 | 68 | |
| CK840972.1 | - | 2 - 3 | 297 | 68 | ||
More Information
Set Membership: |
|

Cox6b2
Genomic Map