Assay Details
| Gene Symbol: Ffar4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047088.1 | NP_001040553.1 | 2 - 3 | 740 | 73 | |
| GenBank mRNA | AB207868.1 | - | 2 - 3 | 740 | 73 | |
More Information
Set Membership: |
|

Ffar4
Genomic Map