Assay Details
| Gene Symbol: LOC367195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047920.1 | NP_001041385.1 | 1 - 2 | 38 | 82 | |
| GenBank mRNA | AY325186.1 | - | 1 - 2 | 38 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY325186.1, NM_001047920.1 transcripts.
Set Membership: |
|

LOC367195
Genomic Map