Assay Details
| Gene Symbol: Prps1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105678.1 | NP_001099148.1 | 1 | 118 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001105678.1 transcripts.
Set Membership: |
|

Prps1l1
Genomic Map