Assay Details
| Gene Symbol: Tktl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106080.1 | NP_001099550.1 | 1 | 1820 | 92 | |
| XM_006253022.2 | XP_006253084.1 | 1 | 1845 | 92 | ||
More Information
Set Membership: |
|

Tktl2
Genomic Map