Assay Details
| Gene Symbol: Rpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031103.1 | NP_112365.1 | 1 - 2 | 28 | 101 | |
| GenBank mRNA | BC058135.1 | - | 1 - 2 | 28 | 6918377 | 101 |
| FQ222282.1 | - | 1 - 2 | 28 | 101 | ||
| J02650.1 | - | 1 - 2 | 28 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058135.1, J02650.1, NM_031103.1 transcripts.
Set Membership: |
|

Rpl19
Genomic Map