Assay Details
| Gene Symbol: Ost4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134690.1 | NP_001128162.1 | 2 | 411 | 93 | |
| NM_001134691.1 | NP_001128163.1 | 3 | 311 | 93 | ||
| GenBank mRNA | BC157814.1 | - | 2 | 389 | 8362186 | 93 |
| BI277067.1 | - | 1 | 160 | 93 | ||
| BI298452.1 | - | 1 | 154 | 93 | ||
| BQ194673.1 | - | 1 | 160 | 93 | ||
| CB706964.1 | - | 3 | 272 | 93 | ||
| Gene Symbol: Agbl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239807.2 | XP_006239869.1 | 15 | 4540 | 93 | |
| XM_006239808.2 | XP_006239870.1 | 16 | 4460 | 93 | ||
| XM_006239810.2 | XP_006239872.1 | 15 | 4343 | 93 | ||
More Information
Set Membership: |
|

Ost4
Genomic Map