Assay Details
| Gene Symbol: Purg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191554.1 | NP_001178483.1 | 1 | 1120 | 96 | |
| XM_006253233.2 | XP_006253295.1 | 1 | 1118 | 96 | ||
More Information
Set Membership: |
|

Purg
Genomic Map