Assay Details
| Gene Symbol: Tra2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057119.1 | NP_476460.1 | 9 | 1616 | 100 | |
| XM_006248536.2 | XP_006248598.1 | 9 | 1660 | 100 | ||
| XM_008768789.1 | XP_008767011.1 | 9 | 5778 | 100 | ||
| XM_008768790.1 | XP_008767012.1 | 10 | 1939 | 100 | ||
| GenBank mRNA | D49708.1 | - | 9 | 1616 | 100 | |
| FQ210017.1 | - | 1 | 201 | 100 | ||
More Information
Set Membership: |
|

Tra2b
Genomic Map