Assay Details
| Gene Symbol: Pom121l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254111.2 | XP_006254173.1 | 1 - 2 | 2369 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pom121l2| Gene Symbol: Pom121l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254111.2 | XP_006254173.1 | 1 - 2 | 2369 | 73 | |
Set Membership: |
|