Assay Details
| Gene Symbol: Insl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252820.1 | XP_006252882.1 | 2 - 3 | 832 | 97 | |
| GenBank mRNA | BC098653.1 | - | 1 - 2 | 1019 | 7386551 | 97 |
More Information
Set Membership: |
|

Insl3
Genomic Map