Assay Details
| Gene Symbol: Klk1c6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003977.1 | NP_001003977.1 | 5 | 685 | 59 | |
| XM_006229110.2 | XP_006229172.1 | 5 | 763 | 59 | ||
More Information
Set Membership: |
|

Klk1c6
Genomic Map