Assay Details
| Gene Symbol: TAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178417.1 | NP_009625.1 | 1 | 1755 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAT1| Gene Symbol: TAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178417.1 | NP_009625.1 | 1 | 1755 | 90 | |
Set Membership: |
|