Assay Details
| Gene Symbol: ATP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001181779.3 | NP_012655.3 | 1 | 1354 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP2| Gene Symbol: ATP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001181779.3 | NP_012655.3 | 1 | 1354 | 107 | |
Set Membership: |
|