Assay Details
| Gene Symbol: SEC72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001182180.1 | NP_013395.1 | 1 | 453 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEC72| Gene Symbol: SEC72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001182180.1 | NP_013395.1 | 1 | 453 | 67 | |
Set Membership: |
|