Assay Details
| Gene Symbol: SEPTIN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135344.5 | XP_003135392.1 | 465 | 83 | ||
| XM_005673867.3 | XP_005673924.1 | 216 | 83 | |||
| XM_005673868.3 | XP_005673925.1 | 442 | 83 | |||
| XM_013986371.2 | XP_013841825.2 | 357 | 83 | |||
| XM_013986372.2 | XP_013841826.2 | 357 | 83 | |||
| XM_013986373.2 | XP_013841827.2 | 356 | 83 | |||
| XM_013986374.2 | XP_013841828.2 | 355 | 83 | |||
| XM_013986377.2 | XP_013841831.1 | 442 | 83 | |||
| XM_021080245.1 | XP_020935904.1 | 465 | 83 | |||
| XM_021080247.1 | XP_020935906.1 | 442 | 83 | |||
| XM_021080248.1 | XP_020935907.1 | 363 | 83 | |||
| XM_021080249.1 | XP_020935908.1 | 442 | 83 | |||
| GenBank mRNA | AK231751.1 | - | 2 - 3 | 179 | 83 | |
| AK237053.1 | - | 1 - 2 | 121 | 83 | ||
| AK240268.1 | - | 1 - 2 | 116 | 83 | ||
| AK349008.1 | - | 1 - 2 | 88 | 83 | ||
| AK390996.1 | - | 2 - 3 | 182 | 83 | ||
| KU358538.1 | - | 1 - 2 | 173 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003135344.5, XM_005673867.3, XM_005673868.3, XM_013986371.2, XM_013986372.2, XM_013986373.2, XM_013986374.2, XM_013986377.2, XM_021080245.1, XM_021080247.1, XM_021080248.1, XM_021080249.1 transcripts.
Set Membership: |
|

SEPTIN6
Genomic Map