Assay Details
| Gene Symbol: ISOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123901.4 | XP_003123949.1 | 364 | 81 | ||
| GenBank mRNA | AK394452.1 | - | 1 - 2 | 319 | 81 | |
| AY610215.1 | - | 1 - 2 | 247 | 81 | ||
More Information
Set Membership: |
|

ISOC1
Genomic Map