Assay Details
| Gene Symbol: ENY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243453.1 | NP_001230382.1 | 3 - 4 | 291 | 86 | |
| NM_001243454.1 | NP_001230383.1 | 3 - 4 | 219 | 86 | ||
| XM_005655342.3 | XP_005655399.1 | 304 | 86 | |||
| GenBank mRNA | AK344028.1 | - | 3 - 4 | 256 | 86 | |
| EW064433.2 | - | 3 - 4 | 195 | 86 | ||
More Information
Set Membership: |
|

ENY2
Genomic Map