Assay Details
| Gene Symbol: DLGAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095868.1 | XP_020951527.1 | 4908 | 57 | ||
| XM_021095869.1 | XP_020951528.1 | 3052 | 57 | |||
| XM_021095870.1 | XP_020951529.1 | 2917 | 57 | |||
| XM_021095871.1 | XP_020951530.1 | 2758 | 57 | |||
More Information
Set Membership: |
|

DLGAP3
Genomic Map