Assay Details
| Gene Symbol: RHOBTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483388.4 | XP_003483436.1 | 2359 | 77 | ||
| XM_021073200.1 | XP_020928859.1 | 1599 | 77 | |||
| XM_021073202.1 | XP_020928861.1 | 1619 | 77 | |||
| XM_021073203.1 | XP_020928862.1 | 1730 | 77 | |||
| XM_021073204.1 | XP_020928863.1 | 3193 | 77 | |||
| XM_021073205.1 | XP_020928864.1 | 2150 | 77 | |||
More Information
Set Membership: |
|

RHOBTB2
Genomic Map