Assay Details
| Gene Symbol: PAFAH1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214250.1 | NP_999415.1 | 10 - 11 | 1668 | 79 | |
| XM_005657017.3 | XP_005657074.1 | 1617 | 79 | |||
| XM_005657018.3 | XP_005657075.1 | 2020 | 79 | |||
| XM_013990003.2 | XP_013845457.1 | 1488 | 79 | |||
| GenBank mRNA | AF319658.1 | - | 10 - 11 | 1668 | 79 | |
| AK231665.1 | - | 6 - 7 | 818 | 79 | ||
More Information
Set Membership: |
|

PAFAH1B1
Genomic Map