Assay Details
| Gene Symbol: CFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025215.1 | NP_001020386.1 | 2 - 3 | 484 | 160 | |
| XM_005656299.3 | XP_005656356.1 | 478 | 160 | |||
| GenBank mRNA | AK240074.1 | - | 2 - 3 | 407 | 160 | |
| AK391778.1 | - | 2 - 3 | 395 | 160 | ||
| AK400614.1 | - | 2 - 3 | 425 | 160 | ||
| AY830125.1 | - | 1 - 2 | 58 | 160 | ||
| DQ058293.1 | - | 2 - 3 | 484 | 160 | ||
| EU561660.1 | - | 2 - 3 | 375 | 160 | ||
| EU561661.1 | - | 2 - 3 | 375 | 160 | ||
More Information
Set Membership: |
|

CFL2
Genomic Map