Assay Details
| Gene Symbol: ARGLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927201.6 | XP_001927236.1 | 863 | 81 | ||
| GenBank mRNA | AK235193.1 | - | 2 - 3 | 807 | 81 | |
More Information
Set Membership: |
|

ARGLU1
Genomic Map