Assay Details
| Gene Symbol: TRIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928904.7 | XP_001928939.3 | 1975 | 98 | ||
| GenBank mRNA | AK349047.1 | - | 4 - 5 | 1623 | 98 | |
More Information
Set Membership: |
|

TRIM8
Genomic Map