Assay Details
| Gene Symbol: YWHAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315726.1 | NP_001302655.1 | 3 - 4 | 586 | 106 | |
| XM_001927228.7 | XP_001927263.3 | 1033 | 106 | |||
| XM_005662949.2 | XP_005663006.1 | 726 | 106 | |||
| XM_021088756.1 | XP_020944415.1 | 1374 | 106 | |||
| XM_021088757.1 | XP_020944416.1 | 839 | 106 | |||
| GenBank mRNA | AK344707.1 | - | 4 - 5 | 677 | 106 | |
| DQ178130.1 | - | 1 - 2 | 101 | 106 | ||
| DQ845179.1 | - | 3 - 4 | 371 | 106 | ||
| JN007378.1 | - | 3 - 4 | 586 | 106 | ||
More Information
Set Membership: |
|

YWHAZ
Genomic Map