Assay Details
| Gene Symbol: RPL10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097477.2 | NP_001090946.2 | 5 - 6 | 548 | 67 | |
| XM_021097852.1 | XP_020953511.1 | 512 | 67 | |||
| GenBank mRNA | AK231620.1 | - | 5 - 6 | 786 | 67 | |
| AK240001.1 | - | 5 - 6 | 523 | 67 | ||
| AK347871.1 | - | 5 - 6 | 518 | 67 | ||
| AK350326.1 | - | 1144 | 67 | |||
| AK392651.1 | - | 5 - 6 | 517 | 67 | ||
| DB788284.1 | - | 5 - 6 | 516 | 67 | ||
| DQ629164.1 | - | 5 - 6 | 515 | 67 | ||
| HX212267.1 | - | 5 - 6 | 549 | 67 | ||
More Information
Set Membership: |
|

RPL10A
Genomic Map