Assay Details
| Gene Symbol: NFIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084124.1 | XP_020939783.1 | 817 | 64 | ||
| XM_021084125.1 | XP_020939784.1 | 986 | 64 | |||
| XM_021084126.1 | XP_020939785.1 | 797 | 64 | |||
| XM_021084127.1 | XP_020939786.1 | 981 | 64 | |||
| XM_021084129.1 | XP_020939788.1 | 823 | 64 | |||
| XM_021084130.1 | XP_020939789.1 | 817 | 64 | |||
| XM_021084131.1 | XP_020939790.1 | 898 | 64 | |||
| XM_021084132.1 | XP_020939791.1 | 796 | 64 | |||
| XM_021084133.1 | XP_020939792.1 | 739 | 64 | |||
| GenBank mRNA | AK232904.1 | - | 5 - 6 | 878 | 64 | |
| AK234160.1 | - | 4 - 5 | 709 | 64 | ||
| AK234856.1 | - | 4 - 5 | 787 | 64 | ||
| AK238540.1 | - | 4 - 5 | 768 | 64 | ||
| AK393640.1 | - | 4 - 5 | 797 | 64 | ||
| AK395170.1 | - | 4 - 5 | 768 | 64 | ||
| AK395253.1 | - | 3 - 4 | 700 | 64 | ||
| AK399039.1 | - | 4 - 5 | 768 | 64 | ||
| AK400854.1 | - | 4 - 5 | 768 | 64 | ||
More Information
Set Membership: |
|

NFIC
Genomic Map