Assay Details
| Gene Symbol: VPS33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098604.1 | NP_001092074.1 | 12 - 13 | 1642 | 122 | |
| XM_013981307.2 | XP_013836761.1 | 1709 | 122 | |||
| GenBank mRNA | AB271950.1 | - | 12 - 13 | 1642 | 122 | |
| AK392660.1 | - | 12 - 13 | 1634 | 122 | ||
More Information
Set Membership: |
|

VPS33A
Genomic Map