Assay Details
| Gene Symbol: CRHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144110.1 | NP_001137582.1 | 8 - 9 | 707 | 65 | |
| XM_021066246.1 | XP_020921905.1 | 1217 | 65 | |||
| GenBank mRNA | AF077185.1 | - | 2 - 3 | 216 | 65 | |
| EU350073.1 | - | 8 - 9 | 707 | 65 | ||
More Information
Set Membership: |
|

CRHR1
Genomic Map