Assay Details
| Gene Symbol: FMOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130105.4 | XP_003130153.1 | 1208 | 62 | ||
| GenBank mRNA | AF159383.1 | - | 1 | 350 | 62 | |
| AK347682.1 | - | 2 | 910 | 62 | ||
More Information
Set Membership: |
|

FMOD
Genomic Map