Assay Details
| Gene Symbol: HTR2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286590.1 | NP_001273519.1 | 6 | 1456 | 84 | |
| XM_021079477.1 | XP_020935136.1 | 1582 | 84 | |||
| GenBank mRNA | AF188614.1 | - | 1 | 253 | 84 | |
| FR720593.1 | - | 6 | 1551 | 84 | ||
| FR720594.1 | - | 6 | 1456 | 84 | ||
More Information
Set Membership: |
|

HTR2C
Genomic Map