Assay Details
| Gene Symbol: CALU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134705.6 | XP_003134753.1 | 968 | 65 | ||
| XM_003484023.4 | XP_003484071.1 | 972 | 65 | |||
| XM_021079308.1 | XP_020934967.1 | 1228 | 65 | |||
| GenBank mRNA | AF257660.1 | - | 5 - 6 | 783 | 65 | |
| AK230838.1 | - | 6 - 7 | 912 | 65 | ||
| AK389826.1 | - | 6 - 7 | 914 | 65 | ||
| AK392887.1 | - | 6 - 7 | 935 | 65 | ||
More Information
Set Membership: |
|

CALU
Genomic Map