Assay Details
| Gene Symbol: TFAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130211.1 | NP_001123683.1 | 3 - 4 | 585 | 67 | |
| GenBank mRNA | AF311909.1 | - | 1 - 2 | 67 | 67 | |
| AK231059.1 | - | 3 - 4 | 418 | 67 | ||
| AK351736.1 | - | 3 - 4 | 398 | 67 | ||
| AK391541.1 | - | 3 - 4 | 400 | 67 | ||
| AY923074.1 | - | 3 - 4 | 585 | 67 | ||
More Information
Set Membership: |
|

TFAM
Genomic Map