Assay Details
| Gene Symbol: PTPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113435.1 | NP_001106906.1 | 7 - 8 | 1035 | 63 | |
| XM_021077276.1 | XP_020932935.1 | 1039 | 63 | |||
| XM_021077277.1 | XP_020932936.1 | 1018 | 63 | |||
| GenBank mRNA | AF455037.1 | - | 5 - 6 | 642 | 63 | |
| EU753192.2 | - | 6 - 7 | 807 | 63 | ||
More Information
Set Membership: |
|

PTPN1
Genomic Map