Assay Details
| Gene Symbol: KCNJ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483648.4 | XP_003483696.2 | 737 | 79 | ||
| GenBank mRNA | AF540391.1 | - | 1 | 346 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KCNJ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483648.4 | XP_003483696.2 | 737 | 79 | ||
| GenBank mRNA | AF540391.1 | - | 1 | 346 | 79 | |
Set Membership: |
|