Assay Details
| Gene Symbol: CSNK2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145377.2 | NP_001138849.1 | 3 - 4 | 313 | 85 | |
| XM_005665790.1 | XP_005665847.1 | 486 | 85 | |||
| GenBank mRNA | AJ301368.1 | - | 1 - 2 | 30 | 85 | |
| AK400043.1 | - | 4 - 5 | 450 | 85 | ||
| EU282347.1 | - | 3 - 4 | 314 | 85 | ||
| HE864475.1 | - | 3 - 4 | 306 | 85 | ||
| HE864476.1 | - | 3 - 4 | 306 | 85 | ||
| HE864480.1 | - | 3 - 4 | 306 | 85 | ||
| HE864481.1 | - | 3 - 4 | 306 | 85 | ||
| HE864482.1 | - | 3 - 4 | 306 | 85 | ||
| X56503.1 | - | 236 | 85 | |||
More Information
Set Membership: |
|

CSNK2B
Genomic Map