Assay Details
| Gene Symbol: ACKR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244095.1 | NP_001231024.1 | 2 | 780 | 104 | |
| GenBank mRNA | AJ585087.1 | - | 1 | 553 | 104 | |
| BX666877.1 | - | 1 | 294 | 104 | ||
| EW281145.2 | - | 1 | 429 | 104 | ||
More Information
Set Membership: |
|

ACKR1
Genomic Map