Assay Details
| Gene Symbol: SSTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190231.1 | NP_001177160.1 | 1 | 753 | 110 | |
| XM_013978119.2 | XP_013833573.1 | 6380 | 110 | |||
| GenBank mRNA | AY138806.2 | - | 1 | 753 | 110 | |
More Information
Set Membership: |
|

SSTR1
Genomic Map