Assay Details
| Gene Symbol: ADORA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243435.2 | NP_001230364.2 | 2 | 1055 | 104 | |
| GenBank mRNA | AK239615.1 | - | 2 | 509 | 104 | |
| AK350215.1 | - | 2 | 493 | 104 | ||
| AY772414.1 | - | 1 | 352 | 104 | ||
| Gene Symbol: TMIGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302771.1 | NP_001289700.1 | 3 | 552 | 104 | |
| GenBank mRNA | CV869081.1 | - | 3 | 550 | 104 | |
More Information
Set Membership: |
|

ADORA3
Genomic Map