Assay Details
| Gene Symbol: SLC27A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278777.1 | NP_001265706.1 | 6 - 7 | 1339 | 67 | |
| GenBank mRNA | AK233289.1 | - | 6 - 7 | 1347 | 67 | |
| AY822678.1 | - | 2 - 3 | 122 | 67 | ||
| JX092264.1 | - | 6 - 7 | 1339 | 67 | ||
More Information
Set Membership: |
|

SLC27A2
Genomic Map