Assay Details
| Gene Symbol: GDF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244310.1 | NP_001231239.1 | 1 | 352 | 93 | |
| XM_021091131.1 | XP_020946790.1 | 418 | 93 | |||
| GenBank mRNA | AY874870.1 | - | 1 | 151 | 93 | |
| HQ657211.2 | - | 1 | 137 | 93 | ||
More Information
Set Membership: |
|

GDF11
Genomic Map