Assay Details
| Gene Symbol: DBH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927211.6 | XP_001927246.3 | 728 | 50 | ||
| GenBank mRNA | DQ139273.1 | - | 3 - 4 | 659 | 50 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DBH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927211.6 | XP_001927246.3 | 728 | 50 | ||
| GenBank mRNA | DQ139273.1 | - | 3 - 4 | 659 | 50 | |
Set Membership: |
|