Assay Details
| Gene Symbol: IL5RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013981906.2 | XP_013837360.1 | 3126 | 74 | ||
| XM_021069274.1 | XP_020924933.1 | 3128 | 74 | |||
| XM_021069275.1 | XP_020924934.1 | 3125 | 74 | |||
| GenBank mRNA | EF491877.1 | - | 5 - 6 | 692 | 74 | |
More Information
Set Membership: |
|

IL5RA
Genomic Map