Assay Details
| Gene Symbol: FFAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278783.1 | NP_001265712.1 | 1 | 757 | 95 | |
| XM_013998289.2 | XP_013853743.1 | 1159 | 95 | |||
| GenBank mRNA | EU366318.1 | - | 1 | 552 | 95 | |
| JQ768802.1 | - | 1 | 757 | 95 | ||
More Information
Set Membership: |
|

FFAR1
Genomic Map