Assay Details
| Gene Symbol: CCT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244180.1 | NP_001231109.1 | 4 - 5 | 326 | 70 | |
| GenBank mRNA | AK238740.1 | - | 4 - 5 | 326 | 70 | |
| L47772.1 | - | 2 - 3 | 121 | 70 | ||
More Information
Set Membership: |
|

CCT3
Genomic Map